accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOH3CCH3H3CH3CONONOH3CCH3

CAS RN 1218790-48-7

IUPAC Name: 1,3-dimethyl-5-{[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methylidene}-1,3-diazinane-2,4,6-trione
Synonyms: 1,3-dimethyl-5-{[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methylidene}-1,3-diazinane-2,4,6-trione
Molecular Weight (g/mol): 370.21
Molecular Formula: C19H23BN2O5
InChi Key: AWWSBYIWDATYAG-UHFFFAOYSA-N
SMILES: CN1C(=O)N(C)C(=O)C(=CC2=CC=C(C=C2)B2OC(C)(C)C(C)(C)O2)C1=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)