Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(1,3-Dimethyl-2,4,6-trioxohexahydropyrimidin-5-ylidenemethyl)benzeneboronic acid pinacol ester, Thermo Scientific™

Catalog No. AAH5112406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5112406 5 g
AAH5112403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5112406 Supplier Thermo Scientific Chemicals Supplier No. H5112406

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1218790-48-7
Molecular Formula C19H23BN2O5
Molecular Weight (g/mol) 370.212
MDL Number MFCD11113040
InChI Key AWWSBYIWDATYAG-UHFFFAOYSA-N
Synonym 1,3-dimethyl-5-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzylidene-pyrimidine-2,4,6-trione, 1,3-dimethyl-5-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methylidene-1,3-diazinane-2,4,6-trione, 4-1,3-dimethyl-2,4,6-trioxohexahydropyrimidin-5-ylidenemethyl benzeneboronic acid pinacol ester, amtb129, 1,3-dimethyl-5-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methylidene pyrimidine-2,4,6 1h,3h,5h-trione, 1-5-benzylidene-1,3-dimethylpyrimidine-2,4,6 1h,3h,5h-trione-4-boronic acid pinacol ester, 5-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzylidene-1,3-dimethylpyrimidine-2,4,6 1h,3h,5h-trione
PubChem CID 46739748
IUPAC Name 1,3-dimethyl-5-[[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methylidene]-1,3-diazinane-2,4,6-trione
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C=C3C(=O)N(C(=O)N(C3=O)C)C

Specifications

Formula Weight 370.21
Quantity 5 g
Chemical Name or Material 4-(1,3-Dimethyl-2,4,6-trioxohexahydropyrimidin-5-ylidenemethyl)benzeneboronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.