accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOH3CCH3H3CH3CFOOCH3F

CAS RN 1218791-32-2

IUPAC Name: methyl 3,5-difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
Synonyms: methyl 3,5-difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
Molecular Weight (g/mol): 298.09
Molecular Formula: C14H17BF2O4
InChi Key: CGADBVGBEFYTMK-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(F)=C(B2OC(C)(C)C(C)(C)O2)C(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results