accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 121905-60-0

IUPAC Name: chloro(3-methylphenyl)magnesium
Synonyms: chloro(3-methylphenyl)magnesium
Molecular Weight (g/mol): 150.89
Molecular Formula: C7H7ClMg
InChi Key: BVYCQPCLCZTSPD-UHFFFAOYSA-M
SMILES: CC1=CC([Mg]Cl)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results