Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

m-Tolylmagnesium chloride, 1M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Catalog No. AC435971000
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC435971000 100 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AC435971000 Supplier Thermo Scientific Chemicals Supplier No. 435971000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 121905-60-0 | Molecular Formula: C7H7ClMg | Molecular Weight: 150.89 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121905-60-0 , 109-99-9
Molecular Formula C7H7ClMg
Molecular Weight (g/mol) 150.89
MDL Number MFCD00238749
InChI Key BVYCQPCLCZTSPD-UHFFFAOYSA-M
Synonym m-tolylmagnesium chloride, m-tolyl magnesium chloride, m-methylphenylmagnesium chloride, bvycqpclcztspd-uhfffaoysa-m, m-tolylmagnesium chloride solution 1.0 m in thf, m-tolylmagnesium chloride solution, 1.0 m in thf
PubChem CID 4384483
SMILES CC1=CC([Mg]Cl)=CC=C1

Specifications

CAS Min % 14.99, 82.64
CAS Max % 17.36, 85.01
Physical Form Solution
Quantity 100 mL
Flash Point −21°C
Solubility Information Solubility in water: reacts
IUPAC Name magnesium;methylbenzene;chloride
Formula Weight 150.89
Concentration or Composition (by Analyte or Components) 0.95 to 1.10M
Packaging AcroSeal™ Glass Bottle
Specific Gravity 0.956
Density 0.9560g/mL
Chemical Name or Material m-Tolylmagnesium chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Reacts violently with water.
May form explosive peroxides.
Suspected of causing cancer.<br/
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.