CAS RN 1219501-75-3
CAS RN 1219501-75-3
IUPAC Name:
5-bromobenzene-1,3-dithiol
Synonyms:
5-bromobenzene-1,3-dithiol
Molecular Weight (g/mol):
221.13
Molecular Formula:
C6H5BrS2
InChi Key:
AHHCKAJHIBGSHJ-UHFFFAOYSA-N
SMILES:
SC1=CC(Br)=CC(S)=C1
1
–
1
of
1
results
1
5-Bromo-1,3-benzenedithiol 98.0+%, TCI America™
CAS: 1219501-75-3 Molecular Formula: C6H5BrS2 Molecular Weight (g/mol): 221.13 MDL Number: MFCD08276291 InChI Key: AHHCKAJHIBGSHJ-UHFFFAOYSA-N Synonym: 1-Bromo-3,5-dimercaptobenzene PubChem CID: 44630533 IUPAC Name: 5-bromobenzene-1,3-dithiol SMILES: SC1=CC(Br)=CC(S)=C1