Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-1,3-benzenedithiol 98.0+%, TCI America™
SDP

Catalog No. B26551G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B26551G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B26551G Supplier TCI America Supplier No. B26551G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1219501-75-3
Molecular Formula C6H5BrS2
Molecular Weight (g/mol) 221.13
MDL Number MFCD08276291
InChI Key AHHCKAJHIBGSHJ-UHFFFAOYSA-N
Synonym 1-Bromo-3,5-dimercaptobenzene
PubChem CID 44630533
IUPAC Name 5-bromobenzene-1,3-dithiol
SMILES SC1=CC(Br)=CC(S)=C1

Specifications

Melting Point 78°C
Color White-Yellow
Quantity 1 g
Formula Weight 221.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-1,3-benzenedithiol
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.