accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OPSOCH3OH3CNOO

CAS RN 122-14-5

IUPAC Name: O,O-dimethyl O-3-methyl-4-nitrophenyl phosphorothioate
Synonyms: fenitrothion MEP
Molecular Weight (g/mol): 277.23
Molecular Formula: C9H12NO5PS
InChi Key: ZNOLGFHPUIJIMJ-UHFFFAOYSA-N
SMILES: COP(=S)(OC)OC1=CC=C(C(C)=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results