accessibility menu, dialog, popup

UserName

Viewing profile as user:
HN

CAS RN 122-39-4

IUPAC Name: N-phenylaniline
Synonyms: diphenylamine caswell no. 398 big dipper shield dpa no-scald dpa 283 diphenylamine (substance) scaldip naugalube 428L deccoscald 282 epa pesticide chemical code 038501
Molecular Weight (g/mol): 169.23
Molecular Formula: C12H11N
InChi Key: DMBHHRLKUKUOEG-UHFFFAOYSA-N
SMILES: N(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (5)
  • (12)
  • (10)
Quantity 
  • (2)
  • (12)
  • (1)
  • (1)
  • (3)
  • (1)
  • (6)
Boiling Point 
  • (16)
  • (9)
Grade 
  • (6)
  • (3)
  • (1)
  • (2)
  • (4)
  • (2)
  • (1)
Physical Form 
  • (3)
  • (2)
  • (2)
  • (4)
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (12)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (5)
  • (1)
  • (15)
  • (1)
  • (1)
  • (1)
  • (5)
  • (12)
  • (10)
  • (2)
  • (12)
  • (1)
  • (1)
  • (3)
  • (1)
  • (6)
  • (16)
  • (9)
  • (6)
  • (3)
  • (1)
  • (2)
  • (4)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (4)
  • (6)
  • (3)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)