Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenylamine (N-Phenylbenzeneamine), Reagents

Catalog No. 01337275
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-275 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-337-275 Supplier Reagents Holdings Llc Supplier No. C2197500100C2
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Diphenylamine (N-Phenylbenzeneamine), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

CAS 122-39-4
Boiling Range 302°C
Melting Point 52.9°C
Color White to Off-White
Boiling Point 302°C
Melting Range 52.9°C
Packaging Glass Bottle
Molecular Formula C12H11N
Quantity 100 g
Specific Gravity 1.16
Synonym N-Phenylbenzeneamine, N-Phenylaniline, N,N-Diphenylamine
InChI Key DMBHHRLKUKUOEG-UHFFFAOYSA-N
SMILES N(C1=CC=CC=C1)C1=CC=CC=C1
IUPAC Name N-phenylaniline
Molecular Weight (g/mol) 169.23
Grade Reagent
Physical Form Solid
Chemical Name or Material Diphenylamine (N-Phenylbenzeneamine)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.