accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOH3COCH3

CAS RN 122-51-0

IUPAC Name: (diethoxymethoxy)ethane
Synonyms: triethyl orthoformate
Molecular Weight (g/mol): 148.2
Molecular Formula: C7H16O3
InChi Key: GKASDNZWUGIAMG-UHFFFAOYSA-N
SMILES: CCOC(OCC)OCC
Boiling Point 
  • (3)
  • (2)
  • (6)
Molecular Weight (g/mol) 
  • (6)
  • (5)
Percent Purity 
  • (2)
  • (9)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (9)
  • (2)
  • (9)
  • (3)
  • (2)
  • (6)
  • (6)
  • (5)
  • (2)
  • (9)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (9)