accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OOOOCH3CH3

CAS RN 122-62-3

IUPAC Name: 1,10-bis(2-ethylhexyl) decanedioate
Synonyms: dioctyl sebacate diethylhexyl sebacate
Molecular Weight (g/mol): 426.68
Molecular Formula: C26H50O4
InChi Key: VJHINFRRDQUWOJ-UHFFFAOYNA-N
SMILES: CCCCC(CC)COC(=O)CCCCCCCCC(=O)OCC(CC)CCCC
Molecular Weight (g/mol) 
  • (4)
  • (9)
Boiling Point 
  • (3)
  • (4)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (4)
  • (1)
  • (2)
  • (6)
  • (4)
  • (9)
  • (3)
  • (4)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (5)
  • (1)