Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Bis(2-ethylhexyl) sebacate

Catalog No. 84822250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
84-822-250ML 250 mL
84-822-1L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 84-822-250ML Supplier Sigma Aldrich Supplier No. 84822250ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Bis(2-ethylhexyl) sebacate is commonly used as a pressure transmitting fluid (ptf) in hydraulically operated devices, pressure measuring instruments and hydraulic pressure balances
  • It is also a plasticizing solvent mediator for preparing poly(vinyl chloride) (pvc) based membranes

Specifications

CAS 122-62-3
Density 0.914 g/mL (at 25°C (literature))
Boiling Point 212°C (1 mmHg)
Molecular Formula C26H50O4
Refractive Index n20/D 1.450 (literature)
Linear Formula [-(CH2)4 CO2CH2CH(C2H5)(CH2)3 CH3]2
MDL Number MFCD00009497
Quantity 250 mL
Synonym Di(2-ethylhexyl) sebacate; Sebacic acid di(2-ethylhexyl) ester; Dioctyl sebacate
Molecular Weight (g/mol) 426.67
Percent Purity ≥97.0% (GC)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.