accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 122-66-7

IUPAC Name: 1,2-diphenylhydrazine
Synonyms: hydrazobenzene
Molecular Weight (g/mol): 184.24
Molecular Formula: C12H12N2
InChi Key: YBQZXXMEJHZYMB-UHFFFAOYSA-N
SMILES: N(NC1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Solvent or Matrix 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)