Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Hydrazobenzene

Catalog No. 501854609
Encompass
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-185-4609 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-185-4609 Supplier Sigma Aldrich Supplier No. 12672125G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Hydrazobenzene
TRUSTED_SUSTAINABILITY

Specifications

CAS 122-66-7
Melting Point 123°C to 126°C (lit.)
Molecular Formula C12H12N2
Linear Formula C6H5NHNHC6H5
MDL Number MFCD00003012
Quantity 250 mg
Synonym N,N-Diphenylhydrazine; N,N-Bianiline; NSC 3510
Molecular Weight (g/mol) 184.24
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.