CAS RN 122-69-0
IUPAC Name:
(2E)-3-phenylprop-2-en-1-yl (2E)-3-phenylprop-2-enoate
Synonyms:
cinnamyl cinnamate
Molecular Weight (g/mol):
264.32
Molecular Formula:
C18H16O2
InChi Key:
NQBWNECTZUOWID-MZXMXVKLSA-N
SMILES:
O=C(OC\C=C\C1=CC=CC=C1)\C=C\C1=CC=CC=C1
1
–
1
of
1
results
1
Cinnamyl Cinnamate 80.0+%, TCI America™
CAS: 122-69-0 Molecular Formula: C18H16O2 Molecular Weight (g/mol): 264.32 MDL Number: MFCD00037826 InChI Key: NQBWNECTZUOWID-MZXMXVKLSA-N Synonym: Cinnamic Acid Cinnamyl Ester PubChem CID: 1550890 IUPAC Name: (2E)-3-phenylprop-2-en-1-yl (2E)-3-phenylprop-2-enoate SMILES: O=C(OC\C=C\C1=CC=CC=C1)\C=C\C1=CC=CC=C1