Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cinnamyl Cinnamate 80.0+%, TCI America™
SDP

Catalog No. C096025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C096025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C096025G Supplier TCI America Supplier No. C096025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 122-69-0
Molecular Formula C18H16O2
Molecular Weight (g/mol) 264.32
MDL Number MFCD00037826
InChI Key NQBWNECTZUOWID-MZXMXVKLSA-N
Synonym Cinnamic Acid Cinnamyl Ester
PubChem CID 1550890
IUPAC Name (2E)-3-phenylprop-2-en-1-yl (2E)-3-phenylprop-2-enoate
SMILES O=C(OC\C=C\C1=CC=CC=C1)\C=C\C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 370°C
Quantity 25 g
Formula Weight 264.32
Percent Purity ≥80.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Cinnamyl Cinnamate
EINECSNumber (3)-3086
RTECSNumber GD8562000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.