accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COO

CAS RN 122-72-5

IUPAC Name: 3-phenylpropyl acetate
Synonyms: hydrocinnamyl acetate
Molecular Weight (g/mol): 178.23
Molecular Formula: C11H14O2
InChi Key: JRJGKUTZNBZHNK-UHFFFAOYSA-N
SMILES: CC(=O)OCCCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results