Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Phenylpropyl Acetate 98.0+%, TCI America™
SDP

Catalog No. A122525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A122525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A122525G Supplier TCI America Supplier No. A122525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 122-72-5
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.231
MDL Number MFCD00026216
InChI Key JRJGKUTZNBZHNK-UHFFFAOYSA-N
Synonym hydrocinnamyl acetate, benzenepropanol, acetate, phenylpropyl acetate, 3-phenyl-1-propyl acetate, 3-acetoxypropyl benzene, 3-phenyl-1-propanol, acetate, 3-acetoxy-1-phenylpropane, 1-acetoxy-3-phenylpropane, benzenepropanol, 1-acetate, benzenepropyl acetate
PubChem CID 31226
ChEBI CHEBI:81257
IUPAC Name 3-phenylpropyl acetate
SMILES CC(=O)OCCCC1=CC=CC=C1

Specifications

Color Colorless
Boiling Point 245°C
Quantity 25 g
Formula Weight 178.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Phenylpropyl Acetate
EINECSNumber (3)-1041&(3)-1045
RTECSNumber UB9000000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.