accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 122-73-6

IUPAC Name: [(3-methylbutoxy)methyl]benzene
Synonyms: benzyl isoamyl ether
Molecular Weight (g/mol): 178.28
Molecular Formula: C12H18O
InChi Key: RXXCIBALSKQCAE-UHFFFAOYSA-N
SMILES: CC(C)CCOCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results