Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl Isoamyl Ether 98.0+%, TCI America™
SDP

Catalog No. I008425ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I008425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I008425ML Supplier TCI America Supplier No. I008425ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 122-73-6
Molecular Formula C12H18O
Molecular Weight (g/mol) 178.275
MDL Number MFCD00048371
InChI Key RXXCIBALSKQCAE-UHFFFAOYSA-N
Synonym Benzyl Isopentyl Ether
PubChem CID 31227
IUPAC Name 3-methylbutoxymethylbenzene
SMILES CC(C)CCOCC1=CC=CC=C1

Specifications

Color Colorless
Boiling Point 236°C
Quantity 25 mL
Formula Weight 178.28
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Benzyl Isoamyl Ether
RTECSNumber KM9620000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.