accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHOCH3OHOHNNH

CAS RN 122-75-8

IUPAC Name: bis(acetic acid); benzyl[2-(benzylamino)ethyl]amine
Synonyms: bis(acetic acid); benzathine
Molecular Weight (g/mol): 360.45
Molecular Formula: C20H28N2O4
InChi Key: MTRNNCLQPVCDLF-UHFFFAOYSA-N
SMILES: CC(O)=O.CC(O)=O.C(CNCC1=CC=CC=C1)NCC1=CC=CC=C1
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)