accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1220219-61-3

IUPAC Name: 2-[2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetonitrile
Synonyms: 2-[2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetonitrile
Molecular Weight (g/mol): 261.1
Molecular Formula: C14H17BFNO2
InChi Key: SAFMDYCIEJEGJQ-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC(F)=C(CC#N)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results