Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Cyanomethyl-3-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals

Catalog No. AAH62103MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH62103MD 250 mg
AAH6210303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH62103MD Supplier Thermo Scientific Chemicals Supplier No. H62103MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1220219-61-3 | Molecular Formula: C14H17BFNO2 | Molecular Weight: 261.103 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1220219-61-3
Molecular Formula C14H17BFNO2
Molecular Weight (g/mol) 261.103
InChI Key SAFMDYCIEJEGJQ-UHFFFAOYSA-N
Synonym 2-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, acmc-209ajh, 2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, 2-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl phenyl acetonitrile, 2-2-fluoro-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile
PubChem CID 54759097
IUPAC Name 2-[2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetonitrile
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)CC#N)F

Specifications

Quantity 250 mg
Formula Weight 261.1
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 4-Cyanomethyl-3-fluorobenzeneboronic acid pinacol ester
Hazard Category H302
Precautionary Statement P264b-P270-P301+P312-P330-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.