accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOHOHOHOHOHKSb

CAS RN 12208-13-8

IUPAC Name: antimony(3+) potassium hexahydroxide
Synonyms: antimony(3+) potassium hexahydroxide
Molecular Weight (g/mol): 262.9
Molecular Formula: H6KO6Sb
InChi Key: IDVSLFDKDQVCAP-UHFFFAOYSA-H
SMILES: [OH-].[OH-].[OH-].[OH-].[OH-].[OH-].[K+].[Sb+3]
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (4)
Grade 
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)