Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Pyroantimonate Solution R, Ricca Chemical
SDP

Catalog No. R6459000500 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R6459000500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R6459000500 Supplier Ricca Chemical Company Supplier No. R6459000500A
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

EP Reagent 1071301/USP for Saccharin Sodium

Specifications

Color Colorless
Grade Laboratory
Packaging Natural Poly Bottle
pH >12
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Potassium Pyroantimonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.