accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 122116-12-5

IUPAC Name: 2-(prop-2-yn-1-yloxy)ethan-1-amine
Synonyms: 2-(prop-2-yn-1-yloxy)ethanamine
Molecular Weight (g/mol): 99.13
Molecular Formula: C5H9NO
InChi Key: MZFRRBQTCNHISM-UHFFFAOYSA-N
SMILES: NCCOCC#C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results