accessibility menu, dialog, popup

UserName

Viewing profile as user:
BSOONHHOOH

CAS RN 1221823-27-3

IUPAC Name: {4-[(cyclohexylmethyl)sulfamoyl]phenyl}boronic acid
Synonyms: 4-(cyclohexylmethylsulfamoyl)phenylboronic acid
Molecular Weight (g/mol): 297.18
Molecular Formula: C13H20BNO4S
InChi Key: JDFPAMLPRYKLRV-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(C=C1)S(=O)(=O)NCC1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results