accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1222689-74-8

IUPAC Name: 9-[(2R,4S,5S)-4-amino-5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-1H-purin-6-one
Synonyms: 9-[(2R,4S,5S)-4-amino-5-(hydroxymethyl)oxolan-2-yl]-1H-purin-6-one
Molecular Weight (g/mol): 251.25
Molecular Formula: C10H13N5O3
InChi Key: CHWLAIBBKPETPZ-YUZWJPFSNA-N
SMILES: N[C@H]1C[C@@H](O[C@@H]1CO)N1C=NC2=C1N=CNC2=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results