Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals 3'-Amino-2',3'-dideoxyinosine, 98%

Catalog No. AAJ6475803
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6475803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6475803 Supplier Thermo Scientific Chemicals Supplier No. J6475803
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1222689-74-8 | C10H13N5O3 | 251.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1222689-74-8
Molecular Formula C10H13N5O3
Molecular Weight (g/mol) 251.25
MDL Number MFCD10566669
InChI Key CHWLAIBBKPETPZ-YUZWJPFSNA-N
Synonym 3'-amino-2',3'-dideoyinosine, 9-2r,4r,5s-4-amino-5-hydroxymethyl oxolan-2-yl-1h-purin-6-one
PubChem CID 71744249
IUPAC Name 9-[(2R,4R,5S)-4-amino-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
SMILES N[C@H]1C[C@@H](O[C@@H]1CO)N1C=NC2=C1N=CNC2=O

Specifications

Color White to Yellow
Quantity 1 g
Solubility Information Soluble in water at 50mg/ml
Formula Weight 251.24
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 3'-Amino-2',3'-dideoxyinosine
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.