accessibility menu, dialog, popup

UserName

Viewing profile as user:
BFFFFPH3N

CAS RN 1223007-01-9

IUPAC Name: 2-{13-phosphapentacyclo[13.8.0.0²,¹¹.0³,⁸.0¹⁸,²³]tricosa-1(23),2,4,6,8,10,15,17,19,21-decaen-13-yl}ethan-1-aminium; tetrafluoroboranuide
Synonyms: 2-{13-phosphapentacyclo[13.8.0.0²,¹¹.0³,⁸.0¹⁸,²³]tricosa-1(23),2,4,6,8,10,15,17,19,21-decaen-13-yl}ethanaminium tetrafluoroborate
Molecular Weight (g/mol): 443.23
Molecular Formula: C24H23BF4NP
InChi Key: AMEHGPFCNMMXTB-UHFFFAOYSA-O
SMILES: F[B-](F)(F)F.[NH3+]CCP1CC2=CC=C3C=CC=CC3=C2C2=C3C=CC=CC3=CC=C2C1
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results