Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-[(11bR)-3H-Dinaphtho[2,1-c:1',2'-e]phosphepin-4(5H)-yl]ethylammonium tetrafluoroborate, 97+%, Thermo Scientific™

Catalog No. AAH60215MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH60215MD 250 mg
AAH6021503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH60215MD Supplier Thermo Scientific Chemicals Supplier No. H60215MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1223007-01-9
Molecular Formula C24H23BF4NP
Molecular Weight (g/mol) 443.232
MDL Number MFCD17018773
InChI Key AMEHGPFCNMMXTB-UHFFFAOYSA-O
Synonym 2-11br-3h-dinaphtho 2,1-c:1',2'-e phosphepin-4 5h-yl ethylammonium tetrafluoroborate, 2-11bs-3h-dinaphtho 2,1-c:1',2'-e phosphepin-4 5h-yl ethylammonium tetrafluoroborate, 2-11br-3,5-dihydro-4h-dinaphtho 2,1-c:1',2'-e phosphepin-4-yl ethaminium tetrafluorborate, 2-11bs-3,5-dihydro-4h-dinaphtho 2,1-c:1',2'-e phosphepin-4-yl ethaminium tetrafluorborate, 2-13-phosphapentacyclo 13.8.0.0_,__.0_,?.0_?,__ tricosa-1 15 ,2,4,6,8,10,16,18,20,22-decaen-13-yl ethanaminium tetrafluoroborate
PubChem CID 72376450
SMILES [B-](F)(F)(F)F.C1C2=C(C3=CC=CC=C3C=C2)C4=C(CP1CC[NH3+])C=CC5=CC=CC=C54

Specifications

Quantity 250 mg
Formula Weight 443.22
Percent Purity ≥97%
Chemical Name or Material 2-[(11bR)-3H-Dinaphtho[2,1-c:1',2'-e]phosphepin-4(5H)-yl]ethylammonium tetrafluoroborate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.