accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 122307-41-9

IUPAC Name: 4-chloro-3-methoxy-2-methylpyridin-1-ium-1-olate
Synonyms: 4-chloro-3-methoxy-2-methylpyridin-1-ium-1-olate
Molecular Weight (g/mol): 173.6
Molecular Formula: C7H8ClNO2
InChi Key: TWXMQDRFBLSXFN-UHFFFAOYSA-N
SMILES: COC1=C(Cl)C=C[N+]([O-])=C1C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results