accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFN

CAS RN 122376-76-5

IUPAC Name: 2-(3,5-difluorophenyl)acetonitrile
Synonyms: 2-(3,5-difluorophenyl)acetonitrile
Molecular Weight (g/mol): 153.13
Molecular Formula: C8H5F2N
InChi Key: OBMYMTSBABEPIB-UHFFFAOYSA-N
SMILES: FC1=CC(CC#N)=CC(F)=C1
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)