Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Difluorobenzyl Cyanide 98.0+%, TCI America™
SDP

Catalog No. D34435G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D34435G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D34435G Supplier TCI America Supplier No. D34435G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 122376-76-5
Molecular Formula C8H5F2N
Molecular Weight (g/mol) 153.132
MDL Number MFCD00061278
InChI Key OBMYMTSBABEPIB-UHFFFAOYSA-N
Synonym 3,5-difluorophenylacetonitrile, 2-3,5-difluorophenyl acetonitrile, 3,5-difluorobenzyl cyanide, 3,5-difluorobenzylcyanide, benzeneacetonitrile, 3,5-difluoro, 3,5-difluorophenyl acetonitrile, 2-3,5-difluorophenyl ethanenitrile, pubchem1588, acmc-1bxc0
PubChem CID 518565
IUPAC Name 2-(3,5-difluorophenyl)acetonitrile
SMILES C1=C(C=C(C=C1F)F)CC#N

Specifications

Melting Point 38°C
Color White-Yellow
Quantity 5 g
UN Number 3439
Formula Weight 153.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Difluorobenzyl Cyanide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.