CAS RN 122384-88-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Amylin, Human
Amylin, Human, CAS 122384-88-7, is a 37-amino acid polypeptide secreted from the β cells of the pancreas. Structurally related to calcitonin. Has anorectic effects in rats.