Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Amylin, Human

Catalog No. 0523254005M Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-232-54005M 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-232-54005M Supplier MilliporeSigma™ Supplier No. 052325400.5MG

Amylin, Human, CAS 122384-88-7, is a 37-amino acid polypeptide secreted from the β cells of the pancreas. Structurally related to calcitonin. Has anorectic effects in rats.

Peptide Sequence: H-Lys-Cys2-Asn-Thr-Ala-Thr-Cys7-Ala-Thr-Gln-Arg-Leu-Ala-Asn-Phe-Leu-Val-His-Ser-Ser-Asn-Asn-Phe-Gly-Ala-Ile-Leu-Ser-Ser-Thr-Asn-Val-Gly-Ser-Asn-Thr-Tyr-NH2 (disulfide bond: 2→7)
TRUSTED_SUSTAINABILITY

Specifications

Color White to Off-white
Formulation Supplied as a trifluoroacetate salt.
Description 97% by HPLC
Target Proteins Has anorectic effects in rats
CAS 122384-88-7
Molecular Formula C165H261N51O55S2
Formula Weight 3903.4g/mol
Solubility 5% Acetic Acid
Content And Storage Protect from moisture
Form Lyophilized
Quantity 0.5 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.