accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1225-13-4

IUPAC Name: (2R,3S)-2,3-diphenylbutanedioic acid
Synonyms: (2R,3S)-2,3-diphenylbutanedioic acid
Molecular Weight (g/mol): 270.28
Molecular Formula: C16H14O4
InChi Key: PVXCQHHWNDJIJP-OKILXGFUSA-N
SMILES: OC(=O)[C@H]([C@H](C(O)=O)C1=CC=CC=C1)C1=CC=CC=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results