Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

meso-2,3-Diphenylsuccinic Acid, TCI America™
SDP

Catalog No. D22091G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D22091G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D22091G Supplier TCI America Supplier No. D22091G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1225-13-4
Molecular Formula C16H14O4
Molecular Weight (g/mol) 270.284
MDL Number MFCD00198082
InChI Key PVXCQHHWNDJIJP-OKILXGFUSA-N
Synonym meso-2,3-Diphenylbutanedioic Acid
PubChem CID 253131
IUPAC Name (2S,3R)-2,3-diphenylbutanedioic acid
SMILES C1=CC=C(C=C1)C(C(C2=CC=CC=C2)C(=O)O)C(=O)O

Specifications

Color White
Quantity 1 g
Formula Weight 270.28
Physical Form Crystalline Powder
Chemical Name or Material meso-2,3-Diphenylsuccinic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.