accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSNSNHSHN

CAS RN 1226494-16-1

IUPAC Name: 6-(methylsulfanyl)-1,2,3,4-tetrahydro-1,3,5-triazine-2,4-dithione
Synonyms: 6-(methylsulfanyl)-1,3-dihydro-1,3,5-triazine-2,4-dithione
Molecular Weight (g/mol): 191.29
Molecular Formula: C4H5N3S3
InChi Key: UVRDIFOZBJEFGI-UHFFFAOYSA-N
SMILES: CSC1=NC(=S)NC(=S)N1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results