Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Si-TMT (=2,4,6-Trimercaptotriazine Silica Gel) (0.2-0.5mmol/g), TCI America™
SDP

Catalog No. S08655G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S08655G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S08655G Supplier TCI America Supplier No. S08655G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1226494-16-1
Molecular Formula C4H5N3S3
Molecular Weight (g/mol) 191.285
InChI Key UVRDIFOZBJEFGI-UHFFFAOYSA-N
Synonym 2,4,6-Trimercaptotriazine Silica Gel
PubChem CID 12210973
IUPAC Name 6-methylsulfanyl-1H-1,3,5-triazine-2,4-dithione
SMILES CSC1=NC(=S)NC(=S)N1

Specifications

Quantity 5 g
Physical Form Crystalline Powder
Chemical Name or Material Si-TMT (=2,4,6-Trimercaptotriazine Silica Gel) (0.2-0.5mmol/g)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.