CAS RN 122651-20-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
phyproof™ 3-O-Acetyl 9,11-dehydro β-boswellic acid, ≥98% (HPLC), MilliporeSigma™ Supelco™
This substance is supplied with certified chromatographic purity. The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG