accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OONNH2O

CAS RN 122684-34-8

IUPAC Name: tert-butyl 3-carbamoylpyrrolidine-1-carboxylate
Synonyms: tert-butyl 3-carbamoylpyrrolidine-1-carboxylate
Molecular Weight (g/mol): 214.27
Molecular Formula: C10H18N2O3
InChi Key: NHDGOVOBEZPXMY-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1CCC(C1)C(N)=O
Molecular Weight (g/mol) 
  • (1)
  • (5)
Quantity 
  • (3)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (5)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)