Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Boc-pyrrolidine-3-carboxamide, 96%, Thermo Scientific™

Catalog No. AAH50081MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH50081MD 250 mg
AAH5008103 1 g
AAH5008106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH50081MD Supplier Thermo Scientific Chemicals Supplier No. H50081MD

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122684-34-8
Molecular Formula C10H18N2O3
Molecular Weight (g/mol) 214.265
MDL Number MFCD04116210
InChI Key NHDGOVOBEZPXMY-UHFFFAOYSA-N
Synonym 3-aminocarbonyl-1-boc-pyrrolidine, 1-boc-3-carbamoylpyrrolidine, 1-boc-pyrrolidine-3-carboxamide, 3-carbamoyl-pyrrolidine-1-carboxylic acid tert-butyl ester, 1-boc-pyrrolidine-3-carboxyamide, tert-butyl 3-aminocarbonyl pyrrolidine-1-carboxylate, acmc-20a2cc, ksc496a4t, 1-boc-3-pyrrolidinecarboxamide, 1-n-boc-pyrrolidine-3-carboxyamide
PubChem CID 4571096
IUPAC Name tert-butyl 3-carbamoylpyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(C1)C(=O)N

Specifications

Quantity 250 mg
Formula Weight 214.27
Percent Purity 96%
Chemical Name or Material 1-Boc-pyrrolidine-3-carboxamide
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.