CAS RN 122738-21-0
CAS RN 122738-21-0
IUPAC Name:
N4,N4,N4',N4'-tetrakis(4-methoxyphenyl)-[1,1'-biphenyl]-4,4'-diamine
Synonyms:
N4,N4,N4',N4'-tetrakis(4-methoxyphenyl)-[1,1'-biphenyl]-4,4'-diamine
Molecular Weight (g/mol):
608.74
Molecular Formula:
C40H36N2O4
InChi Key:
WPUSEOSICYGUEW-UHFFFAOYSA-N
SMILES:
COC1=CC=C(C=C1)N(C1=CC=C(OC)C=C1)C1=CC=C(C=C1)C1=CC=C(C=C1)N(C1=CC=C(OC)C=C1)C1=CC=C(OC)C=C1
1
–
1
of
1
results
1
N,N,N',N'-Tetrakis(4-methoxyphenyl)benzidine 98.0+%, TCI America™

CAS: 122738-21-0 Molecular Formula: C40H36N2O4 Molecular Weight (g/mol): 608.738 MDL Number: MFCD09833415 InChI Key: WPUSEOSICYGUEW-UHFFFAOYSA-N Synonym: MeO-TPD PubChem CID: 23069933 IUPAC Name: 4-[4-(4-methoxy-N-(4-methoxyphenyl)anilino)phenyl]-N,N-bis(4-methoxyphenyl)aniline SMILES: COC1=CC=C(C=C1)N(C2=CC=C(C=C2)C3=CC=C(C=C3)N(C4=CC=C(C=C4)OC)C5=CC=C(C=C5)OC)C6=CC=C(C=C6)OC