Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N,N',N'-Tetrakis(4-methoxyphenyl)benzidine 98.0+%, TCI America™
SDP

Catalog No. T3290200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3290200MG 200 mg
T32901G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T3290200MG Supplier TCI America Supplier No. T3290200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 122738-21-0
Molecular Formula C40H36N2O4
Molecular Weight (g/mol) 608.738
MDL Number MFCD09833415
InChI Key WPUSEOSICYGUEW-UHFFFAOYSA-N
Synonym MeO-TPD
PubChem CID 23069933
IUPAC Name 4-[4-(4-methoxy-N-(4-methoxyphenyl)anilino)phenyl]-N,N-bis(4-methoxyphenyl)aniline
SMILES COC1=CC=C(C=C1)N(C2=CC=C(C=C2)C3=CC=C(C=C3)N(C4=CC=C(C=C4)OC)C5=CC=C(C=C5)OC)C6=CC=C(C=C6)OC

Specifications

Melting Point 192°C
Quantity 200 mg
Formula Weight 608.74
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N,N,N′,N′-Tetrakis(4-methoxyphenyl)benzidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.