accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 123-41-1

IUPAC Name: (2-hydroxyethyl)trimethylazanium hydroxide
Synonyms: sincaline
Molecular Weight (g/mol): 121.18
Molecular Formula: C5H15NO2
InChi Key: KIZQNNOULOCVDM-UHFFFAOYSA-M
SMILES: [OH-].C[N+](C)(C)CCO
Molecular Weight (g/mol) 
  • (8)
  • (3)
Quantity 
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (4)
  • (2)
Grade 
  • (4)
Percent Purity 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (2)
  • (6)
  • (8)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (4)
  • (2)
  • (4)
  • (4)