Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Choline Hydroxide Solution

Catalog No. 292257500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-225-7500ML 500 mL
29-225-7100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 29-225-7500ML Supplier Sigma Aldrich Supplier No. 292257500ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Choline hydroxide solution has been used in the synthesis of choline hexafluoroacetylacetonate that can be used as an extractant along with bis-(trifluoromethylsulfonyl)imide ionic liquid for the homogeneous liquid-liquid extraction of neodymium(III).

Specifications

CAS 123-41-1
Density 1.073 g/mL at 25°C
Molecular Formula C5H15NO2
Linear Formula HOCH2CH2N(CH3)3OH
Refractive Index n20/D 1.4304
MDL Number MFCD00002831
Quantity 500 mL
Molecular Weight (g/mol) 121.18 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.