accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 123-46-6

IUPAC Name: [(hydrazinecarbonyl)methyl]trimethylazanium chloride
Synonyms: [(hydrazinecarbonyl)methyl]trimethylazanium chloride
Molecular Weight (g/mol): 167.64
Molecular Formula: C5H14ClN3O
InChi Key: YSULOORXQBDPCU-UHFFFAOYSA-N
SMILES: [Cl-].C[N+](C)(C)CC(=O)NN
Molecular Weight (g/mol) 
  • (5)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (3)
  • (5)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)