Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Diethyl 2,2-Diethoxyethylphosphonate

Catalog No. D9925025G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D9925025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. D9925025G Supplier Sigma Aldrich Supplier No. D9925025G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reactant for synthesis of:+a,+a-Alkenal derivatives by two-carbon homologationLower rim-phosphonylated rexorcinol calix[4]arenes by condensation reactions+a-Phosphovinyl radicals via a radical trapping sequenceInhibitors of reverse transcriptase via 1,3-dipolar cycloadditionsReactant for Friedel-Crafts reactions

Specifications

CAS 123-46-6
Melting Point 188°C to 192°C
Molecular Formula C5H14ClN3O
Refractive Index n20/D 1.430
Linear Formula [(CH3)3N+CH2CONHNH2]Cl-
MDL Number MFCD00012009
Quantity 25 g
Synonym Diethyl phosphonoacetaldehyde diethyl acetal; NSC 407851; NSC 77101
Molecular Weight (g/mol) 254.26 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.